R&D Systems
Manufacturer Code:FAB4636G100UG
Catalog # FAB4636G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 776123 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human GPR56 Met1-Val342 Accession No. Q9Y653 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: 7-transmembrane protein with no EGF-like N-terminal domains-1; ADGRG1 C-terminal fragment; ADGRG1 CT; ADGRG1 N-terminal fragment; ADGRG1 NT; Adhesion G-protein coupled receptor G1; BFPP EGF-TM7-like G protein-coupled receptor 567-transmembrane protein with no EGF-like N-terminal domains-1 Protein TM7XN1 TM7LN4 TM7LN4G-protein coupled receptor 56 TM7XN1 TM7XN1DKFZp781L1398; G protein-coupled receptor 56; G-protein coupled receptor 56; GPR56 7TM; GPR56 C-terminal fragment; GPR56 CT; GPR56 extracellular subunit; GPR56 N-terminal fragment; GPR56 NT; GPR56 seven-transmembrane subunit; GPR56 subunit alpha; GPR56 subunit beta; Protein TM7XN1; testicular tissue protein Li 77
Gene Aliases: ADGRG1; BFPP; BPPR; GPR56; TM7LN4; TM7XN1; UNQ540/PRO1083
UniProt ID: (Human) Q9Y653
Entrez Gene ID: (Human) 9289
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.