Abnova
Manufacturer Code:PAB26446
Catalog # 89-128-087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 19 amino acids at 1st cytoplasmic domain of human GPR56. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GPR56.
For Research Use Only
Protein Aliases: 7-transmembrane protein with no EGF-like N-terminal domains-1; ADGRG1 C-terminal fragment; ADGRG1 CT; ADGRG1 N-terminal fragment; ADGRG1 NT; Adhesion G-protein coupled receptor G1; BFPP; DKFZp781L1398; G protein-coupled receptor 56; G-protein coupled receptor 56; GPR56 7TM; GPR56 C-terminal fragment; GPR56 CT; GPR56 extracellular subunit; GPR56 N-terminal fragment; GPR56 NT; GPR56 seven-transmembrane subunit; GPR56 subunit alpha; GPR56 subunit beta; Protein TM7XN1; testicular tissue protein Li 77; TM7LN4; TM7XN1
Gene Aliases: ADGRG1; BFPP; BPPR; GPR56; TM7LN4; TM7XN1; UNQ540/PRO1083
UniProt ID: (Human) Q9Y653
Entrez Gene ID: (Human) 9289
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.