Novus Biologicals
Manufacturer Code:NBP19829520UL
Catalog # NBP19829520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is GPR50 - C-terminal region. Peptide sequence ISDIREMQEARTLARARAHARDQAREQDRAHACPAVEETPMNVRNVPLPG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 50; G protein-coupled receptor 50H9Mel1c melatonin-related receptor MGC125342; H9; Melatonin-related receptor
Gene Aliases: GPR50; H9; Mel1c
UniProt ID: (Human) Q13585
Entrez Gene ID: (Human) 9248
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.