Proteintech
Manufacturer Code:215141AP150UL
Catalog # 50-173-3457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GPR50 Fusion Protein Ag16040 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor 50 GPR50 H9 Mel1c Melatonin related receptor; G protein-coupled receptor 50; H9; Melatonin-related receptor
Gene Aliases: GPR50; H9; Mel1c
UniProt ID: (Human) Q13585, (Mouse) O88495
Entrez Gene ID: (Human) 9248, (Mouse) 14765
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.