Invitrogen
Manufacturer Code:PA120419
Catalog # PIPA120419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the 3rd extracellular loop of human G Protein-Coupled Receptor GPR43. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fatty acid receptor 2; FFA2R GPR43; free fatty acid activated receptor 2; Free fatty acid receptor 2; G protein-coupled receptor 43; G-protein coupled receptor 43
Gene Aliases: FFA2; FFA2R; FFAR2; GPCR43; GPR43
UniProt ID: (Human) O15552
Entrez Gene ID: (Human) 2867
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.