Invitrogen
Manufacturer Code:PA5111774
Catalog # PIPA5111774
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 294 - 307 of mouse GPER1 (3rd extracellular loop) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6330420K13Rik CEPR Ceprl Chemoattractant receptor-like 2 chemokine receptor-like 2 CMKRL2 constitutively expressed peptide-like receptor constitutively expressed peptide-like receptor like DRY12 FEG-1 Flow-induced endothelial G-protein coupled receptor 1 G protein-coupled estrogen receptor 1 G protein-coupled receptor 30 GPCR-Br GPER GPER1 Gpr30 GPR41 G-protein coupled estrogen receptor 1 G-protein coupled receptor 30 G-protein coupled receptor 41 heptahelix receptor IL8-related receptor DRY12 LERGU LERGU2 leucine rich protein in GPR30 3'UTR LyGPR Lymphocyte-derived G-protein coupled receptor membrane estrogen receptor mER; Chemoattractant receptor-like 2; chemokine receptor-like 2; constitutively expressed peptide-like receptor; constitutively expressed peptide-like receptor like; FEG-1; Flow-induced endothelial G-protein coupled receptor 1; G protein-coupled estrogen receptor 1; G protein-coupled receptor 30; G-protein coupled estrogen receptor 1; G-protein coupled receptor 30; G-protein coupled receptor 41; GPCR-Br; heptahelix receptor; IL8-related receptor DRY12; leucine rich protein in GPR30 3'UTR; LYGPR; Lymphocyte-derived G-protein coupled receptor; Membrane estrogen receptor; mER
Gene Aliases: 6330420K13Rik; CEPR; Ceprl; CMKRL2; DRY12; FEG-1; GPCR-Br; GPER; GPER1; GPR30; GPR41; LERGU; LERGU2; LyGPR; mER
UniProt ID: (Human) Q99527, (Rat) O08878, (Mouse) Q8BMP4
Entrez Gene ID: (Human) 2852, (Rat) 171104, (Mouse) 76854
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.