Invitrogen
Manufacturer Code:PA5114190
Catalog # PIPA5114190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence: NVFISVHLLQ RTQPGAAPCK QSFRHAHPLT GHIVNLAAFS NSCLNPLIYS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6330420K13Rik CEPR Ceprl Chemoattractant receptor-like 2 chemokine receptor-like 2 CMKRL2 constitutively expressed peptide-like receptor constitutively expressed peptide-like receptor like DRY12 FEG-1 Flow-induced endothelial G-protein coupled receptor 1 G protein-coupled estrogen receptor 1 G protein-coupled receptor 30 GPCR-Br GPER Gper1 GPR30 Gpr41 G-protein coupled estrogen receptor 1 G-protein coupled receptor 30 G-protein coupled receptor 41 heptahelix receptor IL8-related receptor DRY12 LERGU LERGU2 leucine rich protein in GPR30 3&primeUTR LYGPR Lymphocyte-derived G-protein coupled receptor Membrane estrogen receptor mER; Chemoattractant receptor-like 2; chemokine receptor-like 2; constitutively expressed peptide-like receptor; FEG-1; Flow-induced endothelial G-protein coupled receptor 1; G protein-coupled estrogen receptor 1; G protein-coupled receptor 30; G-protein coupled estrogen receptor 1; G-protein coupled receptor 30; GPCR-Br; heptahelix receptor; IL8-related receptor DRY12; leucine rich protein in GPR30 3'UTR; LYGPR; Lymphocyte-derived G-protein coupled receptor; Membrane estrogen receptor; mER
Gene Aliases: CEPR; CMKRL2; DRY12; FEG-1; GPCR-Br; GPER; GPER1; GPR30; LERGU; LERGU2; LyGPR; mER
UniProt ID: (Human) Q99527
Entrez Gene ID: (Human) 2852
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.