Invitrogen
Manufacturer Code:PA570784
Catalog # PIPA570784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human GPR21 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor GPR21; Probable G-protein coupled receptor 21
Gene Aliases: C230004C13Rik; GPR21
UniProt ID: (Human) Q99679, (Mouse) Q8BX79
Entrez Gene ID: (Bovine) 519193, (Pig) 100155256, (Human) 2844, (Rat) 689917, (Mouse) 338346
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.