Invitrogen
Manufacturer Code:PA5100950
Catalog # PIPA5100950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human EBI2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EBI 2 Ebi2 EBV-induced gene 2 EBV-induced G-protein coupled receptor 2 EBV-induced G-protein coupled receptor 2 homolog Epstein-Barr virus induced gene 2 Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupled receptor) Epstein-Barr virus-induced G-protein coupled receptor 2 Epstein-Barr virus-induced G-protein coupled receptor 2 homolog G protein-coupled receptor 183 Gpr183 G-protein coupled receptor 183 RP11-461N23.2; EBI2; EBV-induced G-protein coupled receptor 2; EBV-induced G-protein coupled receptor 2 homolog; Epstein-Barr virus induced gene 2; Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupled receptor); Epstein-Barr virus-induced G-protein coupled receptor 2; Epstein-Barr virus-induced G-protein coupled receptor 2 homolog; G-protein coupled receptor 183
Gene Aliases: EBI2; GPR183
UniProt ID: (Human) P32249, (Mouse) Q3U6B2
Entrez Gene ID: (Human) 1880, (Mouse) 321019
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.