Invitrogen
Manufacturer Code:PA575662
Catalog # PIPA575662
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human GPR182. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrenomedullin receptor; G-protein coupled receptor 182; G-protein coupled receptor 182 GPR182 ADMR; G10D; NOW
Gene Aliases: 7TMR; ADMR; AM-R; AMR; G10-D; G10D; gamrh; Gpcr17; Gpcr22; GPR182; hrhAMR; MB10; NOW
UniProt ID: (Human) O15218
Entrez Gene ID: (Human) 11318, (Mouse) 11536, (Rat) 29307
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.