Invitrogen
Manufacturer Code:PA532813
Catalog # PIPA532813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from C-terminus of human GPR182 / ADMR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 7TMR ADMR AM-R AMR G10D gamrh hrhAMR G-protein coupled receptor 182 adrenomedullin receptor; adrenomedullin receptor; G-protein coupled receptor 182
Gene Aliases: 7TMR; ADMR; AM-R; AMR; G10D; gamrh; GPR182; hrhAMR
UniProt ID: (Human) O15218
Entrez Gene ID: (Human) 11318
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.