Invitrogen
Manufacturer Code:PA5100549
Catalog # PIPA5100549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GPR177 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5031439A09Rik AI173978 AI987742 C1orf139 evenness interrupted homolog EVI G protein-coupled receptor 177 GPR177 Integral membrane protein GPR177 mig-14 MRP Protein evenness interrupted homolog protein wntless homolog Putative NF-kappa-B-activating protein 373 putative NFkB activating protein 373 UNQ85/PRO18667 WLS wntless homolog (Drosophila) wntless Wnt ligand secretion mediator; evenness interrupted homolog; EVI; G protein-coupled receptor 177; Integral membrane protein GPR177; Protein evenness interrupted homolog; Protein wntless homolog; Putative NF-kappa-B-activating protein 373; putative NFkB activating protein 373
Gene Aliases: 5031439A09Rik; AI173978; AI987742; C1orf139; EVI; GPR177; mig-14; MRP; UNQ85/PRO18667; WLS
UniProt ID: (Human) Q5T9L3, (Mouse) Q6DID7, (Rat) Q6P689
Entrez Gene ID: (Human) 79971, (Mouse) 68151, (Rat) 362065
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.