Invitrogen
Manufacturer Code:PA542152
Catalog # PIPA542152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GPR161 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 161; G-protein coupled receptor 161; G-protein coupled receptor RE2
Gene Aliases: Gm208; Gm208Gpr; GPR161; RE2; RGD1563245; vl
UniProt ID: (Human) Q8N6U8, (Mouse) B2RPY5
Entrez Gene ID: (Pig) 100155594, (Horse) 100051574, (Rabbit) 100349764, (Human) 23432, (Dog) 490358, (Rat) 289180, (Guinea pig) 100713924, (Mouse) 240888
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.