Invitrogen
Manufacturer Code:PA5106834
Catalog # PIPA5106834
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GPR152 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A930009H15Rik G protein-coupled receptor 152 G protein-coupled receptor PGR5 Gm673 GPR152 G-protein coupled receptor PGR5 PGR5 Probable G-protein coupled receptor 152 putative G-protein coupled receptor; G protein-coupled receptor PGR5; G-protein coupled receptor PGR5; Probable G-protein coupled receptor 152; putative G-protein coupled receptor
Gene Aliases: A930009H15Rik; Gm673; GPR152; PGR5
UniProt ID: (Human) Q8TDT2, (Mouse) Q8BXS7
Entrez Gene ID: (Human) 390212, (Mouse) 269053
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.