Novus Biologicals
Manufacturer Code:NBP2850060.1ML
Catalog # NB032935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of mouse GPR152 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 152 G protein-coupled receptor PGR5 G-protein coupled receptor PGR5 PGR5MGC148162 probable G-protein coupled receptor 152; G protein-coupled receptor PGR5; G-protein coupled receptor PGR5; Probable G-protein coupled receptor 152
Gene Aliases: GPR152; PGR5
UniProt ID: (Human) Q8TDT2
Entrez Gene ID: (Human) 390212
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.