Novus Biologicals
Manufacturer Code:NBP216717
Catalog # NBP216717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human GPR128. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adhesion G-protein coupled receptor G7; FLJ14454 FLJ16382 FLJ29035 G protein-coupled receptor 128 MGC142011 MGC163260 probable G-protein coupled receptor 128; G protein-coupled receptor 128; G-protein coupled receptor 128
Gene Aliases: ADGRG7; GPR128
UniProt ID: (Human) Q96K78
Entrez Gene ID: (Human) 84873
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.