Abnova
Manufacturer Code:H00166647K
Catalog # 89-206-511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human GPR125 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human GPR125 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Adhesion G protein-coupled receptor A3; G protein-coupled receptor 125; G-protein coupled receptor 125; PGR21 TEM5L; probable G-protein coupled receptor 125
Gene Aliases: ADGRA3; GPR125; PGR21; TEM5L; UNQ556/PRO1113
UniProt ID: (Human) Q8IWK6
Entrez Gene ID: (Human) 166647
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.