Abnova
Manufacturer Code:PAB13307
Catalog # 89-116-556
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus 15 amino acids of human GPR124. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GPR124.
For Research Use Only
Protein Aliases: Adhesion G protein-coupled receptor A2; DKFZp434C211; DKFZp434J0911; FLJ14390; G protein-coupled receptor 124; G-protein coupled receptor 124; KIAA1531; TEM5; Tumor endothelial marker 5
Gene Aliases: ADGRA2; GPR124; KIAA1531; TEM5
UniProt ID: (Human) Q96PE1
Entrez Gene ID: (Human) 25960
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.