Invitrogen
Manufacturer Code:PA599417
Catalog # PIPA599417
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GPR120 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI552415 BMIQ10 FFAR4 free fatty acid receptor 4 G protein-coupled receptor 120 GPCR120 GPR120 GPR129 G-protein coupled receptor 120 G-protein coupled receptor 129 G-protein coupled receptor GT01 G-protein coupled receptor PGR4 G-protein-coupled receptor GT01 GT01 KPG_013 O3FAR1 Omega-3 fatty acid receptor 1 PGR4; Free fatty acid receptor 4; G protein-coupled receptor 120; G-protein coupled receptor 120; G-protein coupled receptor 129; G-protein coupled receptor GT01; G-protein coupled receptor PGR4; G-protein-coupled receptor GT01; Omega-3 fatty acid receptor 1
Gene Aliases: AI552415; BMIQ10; FFAR4; GPR120; GPR129; GT01; KPG_013; O3FAR1; PGR4
UniProt ID: (Human) Q5NUL3, (Mouse) Q7TMA4, (Rat) Q2AC31
Entrez Gene ID: (Human) 338557, (Mouse) 107221, (Rat) 294075
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.