Invitrogen
Manufacturer Code:PA534037
Catalog # PIPA534037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from C-terminus of human O3FAR1 / GPR120. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FFAR4 Free fatty acid receptor 4 GT01 GPR129 GPR120 O3FAR1 PGR4 BMIQ10; Free fatty acid receptor 4; G-protein coupled receptor 120; G-protein coupled receptor 129; G-protein coupled receptor GT01; G-protein coupled receptor PGR4; Omega-3 fatty acid receptor 1
Gene Aliases: BMIQ10; FFAR4; GPR120; GPR129; GT01; O3FAR1; PGR4
UniProt ID: (Human) Q5NUL3
Entrez Gene ID: (Human) 338557
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.