Invitrogen
Manufacturer Code:PA533612
Catalog # PIPA533612
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from internal region of human GPR116. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adhesion G protein-coupled receptor F5; G protein-coupled receptor 116; G-protein coupled receptor 116; GPR116 Ig-Hepta homolog Ig-hepta G protein-coupled receptor 116 KIAA0758 KPG&lowbar001; Ig-Hepta homolog; probable G-protein coupled receptor 116
Gene Aliases: ADGRF5; GPR116; KIAA0758; KPG_001
UniProt ID: (Human) Q8IZF2
Entrez Gene ID: (Human) 221395
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.