Proteintech
Manufacturer Code:140471AP150UL
Catalog # 50-172-9173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GPR116 Fusion Protein Ag5214 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adhesion G protein-coupled receptor F5; G protein coupled receptor 116 GPR116 KIAA0758 KPG&lowbar001; G protein-coupled hepta-helical receptor Ig-Hepta; G protein-coupled receptor 116; G-protein coupled hepta-helical receptor Ig-hepta; G-protein coupled receptor 116; Ig-Hepta homolog; probable G-protein coupled receptor 116
Gene Aliases: 8430401C09Rik; 9330185D23; ADGRF5; AI528491; GPR116; Gprhep; Ig-Hepta; KIAA0758; KPG_001; mKIAA0758
UniProt ID: (Human) Q8IZF2, (Mouse) G5E8Q8, (Rat) Q9WVT0
Entrez Gene ID: (Human) 221395, (Mouse) 224792, (Rat) 245977
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.