R&D Systems
Manufacturer Code:FAB7495N100UG
Catalog # FAB7495N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 909603 |
Immunogen | NS0 mouse myeloma cell line transfected with human GPR101 Met1-Pro508 Accession # NP_473362 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 101 GPCR6 probable G-protein coupled receptor 101; Probable G-protein coupled receptor 101
Gene Aliases: GPCR6; GPR101; PAGH2
UniProt ID: (Human) Q96P66
Entrez Gene ID: (Human) 83550
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.