Invitrogen
Manufacturer Code:PA553733
Catalog # PIPA553733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GPIP137 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activation/proliferation-associated protein 1; caprin 1; Caprin-1; Cell cycle-associated protein 1; Cytoplasmic activation- and proliferation-associated protein 1; cytoplasmic activation/proliferation-associated protein-1; GPI-anchored membrane protein 1; GPI-anchored protein p137; GPI-p137; GPIAP1 GPIP137 M11S1 RNG105 p137GPI GPI-anchored membrane protein 1 GPI-anchored protein p137 GPI-p137 RNA granule protein 105 activation/proliferation-associated protein 1 caprin 1 caprin-1 cell cycle-associated protein 1 cytoplasmic activation- and proliferation-associated protein 1 cytop; Membrane component chromosome 11 surface marker 1; membrane component, chromosome 11, surface marker 1; RNA granule protein 105
Gene Aliases: CAPRIN1; GPIAP1; GPIP137; GRIP137; M11S1; p137GPI; RNG105
UniProt ID: (Human) Q14444
Entrez Gene ID: (Human) 4076
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.