Novus Biologicals
Manufacturer Code:NB11041537SS
Catalog # NB11041537T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to an internal region [within residues 100-170] of mouse GPIHBP1. [Swiss-Prot# Q9D1N2] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endothelial cell LPL transporter; glycosylphosphatidylinositol anchored high density lipoprotein binding protein1 glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1 GPI anchored high density lipoprotein binding protein 1 GPI-anchored HDL-binding protein 1 GPI-HBP1LOC338328 HBP1 High density lipoprotein-binding protein 1; Glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1; GPI anchored high density lipoprotein binding protein 1; GPI-anchored HDL-binding protein 1; GPI-HBP1; High density lipoprotein-binding protein 1
Gene Aliases: GPI-HBP1; GPIHBP1; HBP1; HYPL1D
UniProt ID: (Human) Q8IV16
Entrez Gene ID: (Human) 338328
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.