Novus Biologicals
Manufacturer Code:NBP299379AF405
Catalog # NB085073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human GPI8. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.- GPI transamidase GPI transamidase subunit GPI8 homolog GPI8class K GPI-anchor transamidase hGPI8 MGC22559 phosphatidylinositol glycan anchor biosynthesis class K Phosphatidylinositol-glycan biosynthesis class K protein PIG-K; GPI transamidase; GPI transamidase subunit; GPI-anchor transamidase; GPI8 homolog; hGPI8; phosphatidylinositol glycan anchor biosynthesis, class K; phosphatidylinositol glycan, class K; Phosphatidylinositol-glycan biosynthesis class K protein; PIG-K
Gene Aliases: GPI8; PIGK
UniProt ID: (Human) Q92643
Entrez Gene ID: (Human) 10026
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.