Novus Biologicals
Manufacturer Code:NBP186474
Catalog # NBP186474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:FWSTNIYHLTSFMLENGTSDCNLPENPLFIACGGQQNHTQGSKMQKNDFHRNLTTSLTESVDRNINYTERGVFFSVNSWTPDSMSFIYKALERNIRTMFIGGSQLSQKHVSS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.4.50 Glycoprotein phospholipase D glycosylphosphatidylinositol specific phospholipase D1 glycosylphosphatIdylinositol-specific phospholipase D Glycosyl-phosphatidylinositol-specific phospholipase D GPIPLD GPI-PLD GPIPLDM GPI-specific phospholipase D MGC22590 phosphatidylinositol-glycan-specific phospholipase D phospholipase D phosphatidylinositol-glycan-specific PI-G PLD PIGPLD PIGPLD1; Glycoprotein phospholipase D; Glycosyl-phosphatidylinositol-specific phospholipase D; glycosylphosphatidylinositol specific phospholipase D1, isoform 2; GPI-PLD; GPI-specific phospholipase D; Phosphatidylinositol-glycan-specific phospholipase D; PI-G PLD
Gene Aliases: GPIPLD; GPIPLDM; GPLD1; PIGPLD; PIGPLD1; PLD
UniProt ID: (Human) P80108
Entrez Gene ID: (Human) 2822
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.