Novus Biologicals
Manufacturer Code:NBP30359920UL
Catalog # NB092594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 24-160 of human GPI-PLD/GPLD1 (NP_001494.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.4.50 Glycoprotein phospholipase D glycosylphosphatidylinositol specific phospholipase D1 glycosylphosphatIdylinositol-specific phospholipase D Glycosyl-phosphatidylinositol-specific phospholipase D GPIPLD GPI-PLD GPIPLDM GPI-specific phospholipase D MGC22590 phosphatidylinositol-glycan-specific phospholipase D phospholipase D phosphatidylinositol-glycan-specific PI-G PLD PIGPLD PIGPLD1; Glycoprotein phospholipase D; Glycosyl-phosphatidylinositol-specific phospholipase D; glycosylphosphatidylinositol specific phospholipase D1, isoform 2; GPI-PLD; GPI-specific phospholipase D; Phosphatidylinositol-glycan-specific phospholipase D; PI-G PLD
Gene Aliases: GPIPLD; GPIPLDM; GPLD1; PIGPLD; PIGPLD1; PLD
UniProt ID: (Human) P80108
Entrez Gene ID: (Human) 2822
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.