OriGene
Manufacturer Code:TA501136
Catalog # 50-167-2277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7G10 |
Immunogen | Full length human recombit protein of human GPI produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AMF; AMF GNPI NLK PGI PHI SA-36 SA36; Autocrine motility factor; glucose phosphate isomerase; Glucose-6-phosphate isomerase; GPI; hexose monophosphate isomerase; hexosephosphate isomerase; Neuroleukin; NLK; oxoisomerase; PGI; PHI; Phosphoglucose isomerase; phosphohexomutase; Phosphohexose isomerase; phosphosaccharomutase; SA-36; Sperm antigen 36; sperm antigen-36
Gene Aliases: AMF; GNPI; GPI; NLK; PGI; PHI; SA-36; SA36
UniProt ID: (Human) P06744
Entrez Gene ID: (Human) 2821
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.