Invitrogen
Manufacturer Code:MA535506
Catalog # PIMA535506
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0997 |
Immunogen | A synthesized peptide derived from human Glucose 6 phosphate isomerase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AMF; AMF Autocrine motility factor Glucose-6-phosphate isomerase GNPI Gpi Gpi1 Gpi-1 Gpi-1r Gpi1-r Gpi1s Gpi-1s Gpi1-s Gpi-1t Gpi1-t MF Neuroleukin NK NK/GPI NLK Org PGI PHI Phosphoglucose isomerase Phosphohexose isomerase SA36 SA-36 sperm antigen Sperm antigen 36; Autocrine motility factor; glucose phosphate isomerase; Glucose-6-phosphate isomerase; glucose-6-phosphate isomerase {ECO:0000250|UniProtKB:P06745}; GPI; GPI {ECO:0000250|UniProtKB:P06745}; gpi {ECO:0000312|RGD:2727}; hexose monophosphate isomerase; hexosephosphate isomerase; Neuroleukin; neuroleukin {ECO:0000250|UniProtKB:P06745}; NLK; NLK {ECO:0000250|UniProtKB:P06745}; oxoisomerase; PGI; PGI {ECO:0000250|UniProtKB:P06745}; PHI; PHI {ECO:0000250|UniProtKB:P06745}; Phosphoglucose isomerase; phosphoglucose isomerase {ECO:0000250|UniProtKB:P06745}; phosphohexomutase; Phosphohexose isomerase; phosphohexose isomerase {ECO:0000250|UniProtKB:P06745}; phosphosaccharomutase; SA-36; Sperm antigen 36; sperm antigen-36
Gene Aliases: AMF; GNPI; GPI; Gpi1; NLK; PGI; PHI; SA-36; SA36
UniProt ID: (Human) P06744, (Rat) Q6P6V0
Entrez Gene ID: (Human) 2821, (Rat) 292804
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.