Invitrogen
Manufacturer Code:PA587284
Catalog # PIPA587284
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human GPI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMF; Autocrine motility factor; glucose phosphate isomerase; glucose phosphate isomerase 1 complex; glucose phosphate isomerase 1, regulatory; glucose phosphate isomerase 1, structural; glucose phosphate isomerase 1, temporal; Glucose-6-phosphate isomerase; Glucose-6-phosphate isomerase GPI Autocrine motility factor AMF Neuroleukin NLK Phosphoglucose isomerase PGI Phosphohexose isomerase PHI Sperm antigen 36 SA-36 GPI; glucose-6-phosphate isomerase {ECO:0000250|UniProtKB:P06745}; GPI; GPI {ECO:0000250|UniProtKB:P06745}; gpi {ECO:0000312|RGD:2727}; hexose monophosphate isomerase; hexosephosphate isomerase; maturation factor; Neuroleukin; neuroleukin {ECO:0000250|UniProtKB:P06745}; NLK; NLK {ECO:0000250|UniProtKB:P06745}; oxoisomerase; PGI; PGI {ECO:0000250|UniProtKB:P06745}; PHI; PHI {ECO:0000250|UniProtKB:P06745}; Phosphoglucose isomerase; phosphoglucose isomerase {ECO:0000250|UniProtKB:P06745}; phosphohexomutase; Phosphohexose isomerase; phosphohexose isomerase {ECO:0000250|UniProtKB:P06745}; phosphosaccharomutase; SA-36; Sperm antigen 36; sperm antigen-36
Gene Aliases: AMF; GNPI; GPI; Gpi-1; Gpi-1r; Gpi-1s; Gpi-1t; Gpi1; Gpi1-r; Gpi1-s; Gpi1-t; Gpi1s; MF; NK; NK/GPI; NLK; Org; PGI; PHI; SA-36; SA36
UniProt ID: (Human) P06744, (Rat) Q6P6V0, (Mouse) P06745
Entrez Gene ID: (Human) 2821, (Rat) 292804, (Mouse) 14751
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.