OriGene
Manufacturer Code:TA502218
Catalog # 50-167-3293
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C5 |
Immunogen | Full length human recombit protein of human GPHN produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Domain E; Domain G; GEPH GPH GPHRYN HKPX1 MOCODC; Gephyrin; Molybdopterin adenylyltransferase; Molybdopterin molybdenumtransferase; MPT adenylyltransferase; MPT Mo-transferase
Gene Aliases: GEPH; GPH; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC
UniProt ID: (Human) Q9NQX3
Entrez Gene ID: (Human) 10243
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.