R&D Systems
Manufacturer Code:MAB5534SP
Catalog # MAB5534SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 730737 |
Immunogen | <EM>E. coli</EM>-derived recombinant human GPER Met1-Ser62 Accession # Q99527 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CEPR CEPRG-protein coupled receptor 30 Chemoattractant receptor-like 2 chemokine receptor-like 2 CMKRL2 CMKRL2MGC99678 constitutively expressed peptide-like receptor DRY12 DRY12IL8-related receptor DRY12 FEG-1 FEG-1mER Flow-induced endothelial G-protein coupled receptor 1 G protein-coupled estrogen receptor 1 G protein-coupled receptor 30 GPCR-BR GPR30 GPR30GPCR-Br G-protein coupled estrogen receptor 1 heptahelix receptor LERGU LERGU2 leucine rich protein in GPR30 3'UTR LyGPR Lymphocyte-derived G-protein coupled receptor Membrane estrogen receptor; Chemoattractant receptor-like 2; chemokine receptor-like 2; constitutively expressed peptide-like receptor; FEG-1; Flow-induced endothelial G-protein coupled receptor 1; G protein-coupled estrogen receptor 1; G protein-coupled receptor 30; G-protein coupled estrogen receptor 1; G-protein coupled receptor 30; GPCR-Br; heptahelix receptor; IL8-related receptor DRY12; leucine rich protein in GPR30 3'UTR; LYGPR; Lymphocyte-derived G-protein coupled receptor; Membrane estrogen receptor; mER
Gene Aliases: CEPR; CMKRL2; DRY12; FEG-1; GPCR-Br; GPER; GPER1; GPR30; LERGU; LERGU2; LyGPR; mER
UniProt ID: (Human) Q99527
Entrez Gene ID: (Human) 2852
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.