Abnova
Manufacturer Code:PAB26364
Catalog # 89-128-004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acids at N-terminal extracellular domain of human GPER. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GPER.
For Research Use Only
Protein Aliases: CEPR; Chemoattractant receptor-like 2; chemokine receptor-like 2; CMKRL2; constitutively expressed peptide-like receptor; DRY12; FEG-1; Flow-induced endothelial G-protein coupled receptor 1; G protein-coupled estrogen receptor 1; G protein-coupled receptor 30; G-protein coupled estrogen receptor 1; G-protein coupled receptor 30; GPCR-Br; GPR30; heptahelix receptor; IL8-related receptor DRY12; LERGU; LERGU2; leucine rich protein in GPR30 3'UTR; LYGPR; Lymphocyte-derived G-protein coupled receptor; Membrane estrogen receptor; mER; MGC99678
Gene Aliases: CEPR; CMKRL2; DRY12; FEG-1; GPCR-Br; GPER; GPER1; GPR30; LERGU; LERGU2; LyGPR; mER
UniProt ID: (Human) Q99527
Entrez Gene ID: (Human) 2852
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.