Invitrogen
Manufacturer Code:PA552502
Catalog # PIPA552502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GPD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDH2 GPDM mGPDH GPD-M GPDH-M glycerol-3-phosphate dehydrogenase mitochondrial mitochondrial glycerophosphate dehydrogenase mtGPD; glycerol-3-phosphate dehydrogenase 2 (mitochondrial); Glycerol-3-phosphate dehydrogenase, mitochondrial; GPD-M; GPDH-M; mGDH; mitochondrial glycerophosphate dehydrogenase; mitohondrial glycerophosphate dehydrogenase gene; mtGPD; testicular tissue protein Li 76
Gene Aliases: GDH2; GPD2; GPDM; mGPDH
UniProt ID: (Human) P43304
Entrez Gene ID: (Human) 2820
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.