Invitrogen
Manufacturer Code:PA5100148
Catalog # PIPA5100148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GPD2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA408484 AI448216 AU021455 AW494132 FAD-linked glycerol-3-phosphate dehydrogenase GDH2 Gdm1 glycerol phosphate dehydrogenase 1 mitochondrial glycerol phosphate dehydrogenase 2 mitochondrial Glycerol-3-phosophate dehydrogenase 2 (mitochondrial) glycerol-3-phosphate dehydrogenase 2 glycerol-3-phosphate dehydrogenase 2 (mitochondrial) glycerol-3-phosphate dehydrogenase 2 mitochondrial glycerol-3-phosphate dehydrogenase mitochondrial Gpd2 GPDH GPDH-M GPDM GPD-M mGPDH mitochondrial glycerol-3-phosphate dehydrogenase mitochondrial glycerophosphate dehydrogenase mtGPD mtGPDH mtGPDH gene promoter region and alternative transcripts Protein TISP38 testicular tissue protein Li 76 TISP38; FAD-linked glycerol-3-phosphate dehydrogenase; glycerol phosphate dehydrogenase 1, mitochondrial; Glycerol-3-phosophate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2, mitochondrial; Glycerol-3-phosphate dehydrogenase, mitochondrial; GPD-M; GPDH-M; mGDH; mitochondrial glycerophosphate dehydrogenase; mitohondrial glycerophosphate dehydrogenase gene; mtGPD; mtGPDH gene, promoter region and alternative transcripts; Protein TISP38; testicular tissue protein Li 76
Gene Aliases: AA408484; AI448216; AU021455; AW494132; GDH2; Gdm1; Gpd-m; GPD2; GPDH; Gpdh-m; GPDM; mGPDH; mtGPDH; TISP38
UniProt ID: (Human) P43304, (Mouse) Q64521, (Rat) P35571
Entrez Gene ID: (Human) 2820, (Mouse) 14571, (Rat) 25062
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.