Novus Biologicals
Manufacturer Code:NBP259439100UL
Catalog # NBP259439X
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1400 |
Immunogen | Recombinant human GPD1L (1-351aa) purified from E. coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 1.1.1 EC 1.1.1.8 glycerol-3-phosphate dehydrogenase 1-like glycerol-3-phosphate dehydrogenase 1-like protein KIAA0089GPD1-L; Glycerol-3-phosphate dehydrogenase 1-like protein; GPD1-L
Gene Aliases: GPD1-L; GPD1L; KIAA0089
UniProt ID: (Human) Q8N335
Entrez Gene ID: (Human) 23171
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.