Invitrogen
Manufacturer Code:PA5114046
Catalog # PIPA5114046
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human GPD1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2210409H23Rik D9Ertd660e glycerol-3-phosphate dehydrogenase 1-like Glycerol-3-phosphate dehydrogenase 1-like protein GPD1L GPD1-L Kiaa0089 RGD1560123; Glycerol-3-phosphate dehydrogenase 1-like protein; GPD1-L
Gene Aliases: GPD1-L; GPD1L; KIAA0089
UniProt ID: (Human) Q8N335
Entrez Gene ID: (Human) 23171
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.