Novus Biologicals
Manufacturer Code:NBP30469620UL
Catalog # NB094443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-351 of human GPD1L (NP_055956.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.1.1 EC 1.1.1.8 glycerol-3-phosphate dehydrogenase 1-like glycerol-3-phosphate dehydrogenase 1-like protein KIAA0089GPD1-L; Glycerol-3-phosphate dehydrogenase 1-like protein; GPD1-L
Gene Aliases: GPD1-L; GPD1L; KIAA0089
UniProt ID: (Human) Q8N335
Entrez Gene ID: (Human) 23171
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.