Proteintech
Manufacturer Code:172631AP150UL
Catalog # 50-173-1476
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GPD1L Fusion Protein Ag10977 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycerol-3-phosphate dehydrogenase 1-like protein; GPD1 L GPD1L KIAA0089; GPD1-L
Gene Aliases: 2210409H23Rik; D9Ertd660e; GPD1-L; GPD1L; KIAA0089; RGD1560123
UniProt ID: (Human) Q8N335, (Mouse) Q3ULJ0
Entrez Gene ID: (Human) 23171, (Mouse) 333433, (Rat) 363159
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.