Invitrogen
Manufacturer Code:PA527971
Catalog # PIPA527971
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 37 and 296 of Human GPD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glycerol 3-phosphate dehydrogenase; glycerol phosphate dehydrogenase 1, cytoplasmic adult; glycerol-3-phosphate dehydrogenase 1 (soluble); Glycerol-3-phosphate dehydrogenase [NAD(+)], cytoplasmic; glycerol-3-phosphate dehydrogenase [NAD+], cytoplasmic; glycerolphosphate dehydrogenase 1, cytoplasmic adult; glycerophosphate dehydrogenase; GPD-C; GPD-C GPDH-C glycerol-3-phosphate dehydrogenase &lsqbNAD+] cytoplasmic glycerol-3-phosphate dehydrogenase soluble; GPDH-C
Gene Aliases: AI747587; Gdc-1; Gdc1; GPD-C; GPD1; Gpd3; GPDH; GPDH-C; HTGTI; Kiaa4010; mKIAA4010
UniProt ID: (Human) P21695, (Mouse) P13707, (Rat) O35077
Entrez Gene ID: (Human) 2819, (Mouse) 14555, (Rat) 60666
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.