Invitrogen
Manufacturer Code:PA565346
Catalog # PIPA565346
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human GPCPD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endometrial differential 3; GDE5 KIAA1434; glycerophosphocholine phosphodiesterase GDE1 homolog; Glycerophosphocholine phosphodiesterase GPCPD1; Glycerophosphodiester phosphodiesterase 5; preimplantation protein 4; putative glycerophosphocholine phosphodiesterase GPCPD1; putative glycerophosphodiester phosphodiesterase 5
Gene Aliases: EDI3; GDE5; GDPD6; GPCPD1; KIAA1434; PREI4
UniProt ID: (Human) Q9NPB8
Entrez Gene ID: (Human) 56261
Molecular Function: hydrolase lipase phosphodiesterase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.