Novus Biologicals
Manufacturer Code:NBP255271
Catalog # NBP255271
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NAVALLPENDTGESMLWKATIVLSRGVSVQYRYFKGYFLEPKTIGGPCQVIVHKWETHLQPRSITPLESEIIIDDGQFGIHNGVETLDSGWLTCQTEIRLRLHYSEKPPVSITKKKLKKSRFRVKLTLEGLEEDDDDRVSPTVLHKMS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1 FLJ11085 GDE5PREI4 GDPD6 glycerophosphocholine phosphodiesterase GDE1 homolog (S. cerevisiae) Glycerophosphodiester phosphodiesterase 5 KIAA1434preimplantation protein 4 MGC26147 putative glycerophosphocholine phosphodiesterase GPCPD1 putative glycerophosphodiester phosphodiesterase 5; endometrial differential 3; glycerophosphocholine phosphodiesterase GDE1 homolog; Glycerophosphocholine phosphodiesterase GPCPD1; Glycerophosphodiester phosphodiesterase 5; preimplantation protein 4; putative glycerophosphocholine phosphodiesterase GPCPD1; putative glycerophosphodiester phosphodiesterase 5
Gene Aliases: EDI3; GDE5; GDPD6; GPCPD1; KIAA1434; PREI4
UniProt ID: (Human) Q9NPB8
Entrez Gene ID: (Human) 56261
Molecular Function:
hydrolase
lipase
phosphodiesterase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.