Invitrogen
Manufacturer Code:PA589290
Catalog # PIPA589290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 544-843 of human PYGB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain glycogen phosphorylase; glycogen phosphorylase B; Glycogen phosphorylase, brain form
Gene Aliases: GLYPHOA; GPBB; PYGB
UniProt ID: (Human) P11216, (Mouse) Q8CI94, (Rat) P53534
Entrez Gene ID: (Human) 5834, (Mouse) 110078, (Rat) 25739
Molecular Function: phosphorylase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.