Invitrogen
Manufacturer Code:PA5115315
Catalog # PIPA5115315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GPAT-2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase GPAT2; A530057A03Rik cancer/testis antigen 123 CT123 Glycerol-3-phosphate acyltransferase 2 mitochondrial glycerol-3-phosphate acyltransferase-like 1 Gm116 GPAT2 GPAT-2 RGD1304904 xGPAT1; cancer/testis antigen 123; Glycerol-3-phosphate acyltransferase 2, mitochondrial; glycerol-3-phosphate acyltransferase-like 1; GPAT-2; xGPAT1
Gene Aliases: A530057A03Rik; CT123; Gm116; GPAT2; RGD1304904; xGPAT1
UniProt ID: (Human) Q6NUI2, (Mouse) Q14DK4, (Rat) D3ZI76
Entrez Gene ID: (Human) 150763, (Mouse) 215456, (Rat) 296130
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.