Invitrogen
Manufacturer Code:PA520524
Catalog # PIPA520524
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the carboxy terminus of the human GPAT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glycerol 3-phosphate acyltransferase, mitochondrial; Glycerol-3-phosphate acyltransferase 1, mitochondrial; GPAT; GPAT-1; P90; RP11-426E5.2 GPAT GPAT1 GPAT-1 glycerol 3-phosphate acyltransferase mitochondrial glycerol-3-phosphate acyltransferase 1 mitochondrial
Gene Aliases: GPAM; GPAT; GPAT1; KIAA1560
UniProt ID: (Human) Q9HCL2, (Mouse) Q61586, (Rat) P97564
Entrez Gene ID: (Human) 57678, (Mouse) 14732, (Rat) 29653
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.