Invitrogen
Manufacturer Code:PA542863
Catalog # PIPA542863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human GPAA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anchor attachment protein 1; anchor attachment protein 1 (Gaa1p, yeast) homolog; GAA1 protein homolog; glycophosphatidylinositol anchor attachment 1; glycosylphosphatidylinositol anchor attachment 1 (GPAA1); Glycosylphosphatidylinositol anchor attachment 1 protein; glycosylphosphatidylinositol anchor attachment protein 1 homolog; Glycosylphosphatidylinositol anchor attachment protein 1 homolog (yeast) GAA1 hGAA1; GPAA1P anchor attachment protein 1 homolog; GPI anchor attachment protein 1; GPI transamidase subunit; hGAA1; mGAA1
Gene Aliases: C80044; GAA1; GPAA1; hGAA1; mGAA1
UniProt ID: (Human) O43292, (Mouse) Q9WTK3
Entrez Gene ID: (Bovine) 513395, (Human) 8733, (Dog) 475120, (Horse) 100063382, (Rabbit) 100341924, (Mouse) 14731, (Rat) 300046, (Guinea pig) 100713167
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.