Invitrogen
Manufacturer Code:PA577476
Catalog # PIPA577476
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)DTLYMVHMAAYTEEG corresponding to amino acid residues 584-598 of rat GP130 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD130; gp130; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6R-beta; IL-6RB; Interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; Membrane glycoprotein 130; Oncostatin-M receptor subunit alpha
Gene Aliases: 5133400A03Rik; AA389424; Ac1055; BB405851; CD130; D13Ertd699e; Gp130; Il-6rb; Il6st
UniProt ID: (Mouse) Q00560
Entrez Gene ID: (Rat) 25205, (Mouse) 16195
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.