Invitrogen
Manufacturer Code:MA530909
Catalog # PIMA530909
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9D7A9 |
Immunogen | Recombit Rat gp130/CD130/IL6ST protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD130 CDw130 GP130 GP130-RAPS IL6R-beta IL6ST; IL-6 receptor subunit beta; IL-6R subunit beta; interleukin-6 receptor subunit beta; interleukin-6 signal transducer; membrane glycoprotein 130; oncostatin-M receptor subunit alpha
Gene Aliases: Ac1055; Gp130; Il-6rb
Entrez Gene ID: (Rat) 25205
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.