Invitrogen
Manufacturer Code:MA536040
Catalog # PIMA536040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 022 |
Immunogen | Recombinant Mouse IL6ST/gp130/CD130 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 5133400A03Rik AA389424 Ac1055 BB405851 CD130 CD130 antigen CDw130 D13Ertd699e EGK_16499 gp130 gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form gp130 oncostatin M receptor GP130-RAPS IL-6 IL-6 receptor subunit beta IL-6R subunit beta Il-6rb IL-6R-beta IL6R-beta IL6ST interleukin 6 signal transducer interleukin 6 signal transducer (gp130 oncostatin M receptor) interleukin 6 signal transducer receptor interleukin receptor beta chain Interleukin-6 receptor subunit beta Interleukin-6 signal transducer Membrane glycoprotein 130 membrane glycoprotein gp130 Oncostatin-M receptor subunit alpha; CD130; gp130; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6R-beta; IL-6RB; Interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; Membrane glycoprotein 130; Oncostatin-M receptor subunit alpha
Gene Aliases: 5133400A03Rik; AA389424; BB405851; CD130; D13Ertd699e; gp130; Il6st
UniProt ID: (Mouse) Q00560
Entrez Gene ID: (Mouse) 16195
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.